Novus Biologicals
Manufacturer Code:NBP289956500UG
Catalog # NB021379
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | R001 |
Immunogen | Recombinant Human TRANCE/TNFSF11/RANK L Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: CD254; CD254 CD254 antigen OPGLOPTB2 Osteoclast differentiation factor Osteoprotegerin ligand RANKLreceptor activator of nuclear factor kappa B ligand Receptor activator of nuclear factor kappa-B ligand sOdf TNF-related activation-induced cytokine TRANCEODFhRANKL2 tumor necrosis factor (ligand) superfamily member 11 tumor necrosis factor ligand superfamily member 11; ODF; OPGL; Osteoclast differentiation factor; Osteoprotegerin ligand; RANKL; receptor activator of nuclear factor kappa B ligand; Receptor activator of nuclear factor kappa-B ligand; TNF-related activation-induced cytokine; TRANCE; tumor necrosis factor (ligand) superfamily member 11; tumor necrosis factor (ligand) superfamily, member 11; tumor necrosis factor ligand 6B; Tumor necrosis factor ligand superfamily member 11; Tumor necrosis factor ligand superfamily member 11, membrane form; Tumor necrosis factor ligand superfamily member 11, soluble form
Gene Aliases: CD254; hRANKL2; ODF; OPGL; OPTB2; RANKL; sOdf; TNFSF11; TNLG6B; TRANCE
UniProt ID: (Human) O14788
Entrez Gene ID: (Human) 8600
Molecular Function: cytokine signaling molecule tumor necrosis factor family member
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.