Invitrogen
Manufacturer Code:PA5111544
Catalog # PIPA5111544
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human TRAK2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2900022D04Rik 4733401O11Rik AI480836 Als2cr3 Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein homolog AW046271 CALS-C EMBL AAH60681.1 GABA-A receptor-interacting factor 1 Grif1 GRIF-1 KIAA0549 MILT2 mKIAA0549 O-GlcNAc transferase-interacting protein of 98 kDa Oip98 Trafficking kinesin-binding protein 2 Trak2; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 3; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein; gamma-aminobutyric acid(A) receptor-interacting factor; milton homolog 2; O-linked N-acetylglucosamine transferase interacting protein 98; Trafficking kinesin-binding protein 2; trafficking protein, kinesin binding 2
Gene Aliases: ALS2CR3; CALS-C; GRIF-1; GRIF1; KIAA0549; MILT2; OIP98; TRAK2
UniProt ID: (Human) O60296
Entrez Gene ID: (Human) 66008
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.