Abnova
Manufacturer Code:PAB20850
Catalog # 89-122-347
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human TRAK2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant TRAK2.
For Research Use Only
Protein Aliases: ALS2CR3; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 3; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein; CALS-C; gamma-aminobutyric acid(A) receptor-interacting factor; GRIF-1; GRIF1; KIAA0549; milton homolog 2; O-linked N-acetylglucosamine transferase interacting protein 98; OIP98; Trafficking kinesin-binding protein 2; trafficking protein, kinesin binding 2
Gene Aliases: ALS2CR3; CALS-C; GRIF-1; GRIF1; KIAA0549; MILT2; OIP98; TRAK2
UniProt ID: (Human) O60296
Entrez Gene ID: (Human) 66008
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.