Proteintech
Manufacturer Code:137701AP150UL
Catalog # 50-172-8996
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TRAK2 Fusion Protein Ag4753 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ALS2CR3 CALS C GRIF 1 GRIF1 KIAA0549 OIP98 TRAK2; amyotrophic lateral sclerosis 2 (juvenile) chromosome region candidate 3; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 3; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 3 homolog; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein homolog; GABA-A receptor interacting factor-1; GABA-A receptor-interacting factor 1; gamma-aminobutyric acid(A) receptor-interacting factor; GRIF-1; milton homolog 2; O-GlcNAc transferase-interacting protein of 98 kDa; O-linked N-acetylglucosamine transferase interacting protein 98; Trafficking kinesin-binding protein 2; trafficking protein, kinesin binding 2
Gene Aliases: 2900022D04Rik; 4733401O11Rik; AI480836; ALS2CR3; AW046271; CALS-C; GRIF-1; GRIF1; KIAA0549; MILT2; mKIAA0549; OIP98; TRAK2
UniProt ID: (Human) O60296, (Rat) Q8R2H7
Entrez Gene ID: (Human) 66008, (Mouse) 70827, (Rat) 171086
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.