Novus Biologicals
Manufacturer Code:NBP231343
Catalog # NBP231343
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | DR-4-02 |
Immunogen | Fusion protein (Human). Fusion protein containing the extracellular part of DR4 and the constant part of the heavy chain of the human IgG1 (NM_003844.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: APO2 CD261 CD261 antigen cytotoxic TRAIL receptor Death receptor 4 DR4 DR4 TRAIL receptor 1 TNF-related apoptosis inducing ligand receptor 1 TNF-related apoptosis-inducing ligand receptor 1 TRAIL-R1 TRAILR-1 TRAILR1MGC9365 tumor necrosis factor receptor superfamily member 10A tumor necrosis factor receptor superfamily member 10a; CD261; cytotoxic TRAIL receptor; Death receptor 4; TNF-related apoptosis-inducing ligand receptor 1; TRAIL receptor 1; TRAIL-R1; Tumor necrosis factor receptor superfamily member 10A; tumor necrosis factor receptor superfamily member 10a variant 2; tumor necrosis factor receptor superfamily, member 10a
Gene Aliases: APO2; CD261; DR4; TNFRSF10A; TRAILR-1; TRAILR1
UniProt ID: (Human) O00220
Entrez Gene ID: (Human) 8797
Molecular Function: cytokine receptor receptor tumor necrosis factor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.