Novus Biologicals
Manufacturer Code:NBP29301920UL
Catalog # NB027934
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 386-625 of human TRAF3IP1 (NP_001132962.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp434F124 Interleukin-13 receptor alpha 1-binding protein 1 Microtubule-interacting protein associated with TRAF3 microtubule-interacting protein that associates with TRAF3 MIPT3interleukin 13 receptor alpha 1-binding protein-1 MIP-T3microtubule interacting protein that associates with TRAF3 TNF receptor-associated factor 3 interacting protein 1 TRAF3-interacting protein 1; interleukin 13 receptor alpha 1-binding protein-1; Interleukin-13 receptor alpha 1-binding protein 1; Intraflagellar transport protein 54 homolog; microtubule interacting protein that associates with TRAF3; Microtubule-interacting protein associated with TRAF3; microtubule-interacting protein that associates with TRAF3; MIP-T3; TNF receptor-associated factor 3 interacting protein 1; TRAF3-interacting protein 1
Gene Aliases: IFT54; MIP-T3; MIPT3; SLSN9; TRAF3IP1
UniProt ID: (Human) Q8TDR0
Entrez Gene ID: (Human) 26146
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.