Proteintech
Manufacturer Code:144041AP150UL
Catalog # 50-172-9425
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TRAF3IP1 Fusion Protein Ag5677 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: interleukin 13 receptor alpha 1-binding protein-1; Interleukin-13 receptor alpha 1-binding protein 1; Intraflagellar transport protein 54 homolog; microtubule interacting protein that associates with TRAF3; Microtubule-interacting protein associated with TRAF3; microtubule-interacting protein that associates with TRAF3; MIP T3 MIPT3 TRAF3 interacting protein 1 TRAF3IP1; MIP-T3; TNF receptor-associated factor 3 interacting protein 1; TRAF3-interacting protein 1
Gene Aliases: 3930402D05Rik; AU041749; IFT54; MIP-T3; MIPT3; SLSN9; TRAF3IP1
UniProt ID: (Human) Q8TDR0, (Rat) Q5XIN3, (Mouse) Q149C2
Entrez Gene ID: (Human) 26146, (Rat) 363286, (Mouse) 74019
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.