Abnova
Manufacturer Code:PAB12955
Catalog # 89-116-216
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus 15 amino acids of human TRAF3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of TRAF3.
For Research Use Only
Protein Aliases: CAP-1; CD40 associated protein 1; CD40 binding protein; CD40 receptor associated factor 1; CD40 receptor-associated factor 1; CD40-binding protein; CD40BP; CRAF1; LAP1; LMP1-associated protein 1; RING-type E3 ubiquitin transferase TRAF3; TNF receptor-associated factor 3
Gene Aliases: CAP-1; CAP1; CD40bp; CRAF1; IIAE5; LAP1; TRAF3
UniProt ID: (Human) Q13114
Entrez Gene ID: (Human) 7187
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.