Invitrogen
Manufacturer Code:711288
Catalog # PI711288
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Oligoclonal |
Type | Antibody |
Clone | 12HCLC |
Immunogen | Peptides corresponding to Human TRAF2 (aa 483-501 38-53) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage,avoiding freeze/thaw cycles.,store at -20°C |
Rabbit Recombinant Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase TRAF2; RING-type E3 ubiquitin transferase TRAF2; TNF Receptor Associated Factor 2; TNF receptor-associated factor 2; tumor necrosis factor type 2 receptor associated protein 3; Tumor necrosis factor type 2 receptor-associated protein 3
Gene Aliases: MGC:45012; TRAF2; TRAP; TRAP3
UniProt ID: (Human) Q12933
Entrez Gene ID: (Human) 7186
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.