Invitrogen
Manufacturer Code:PA517500
Catalog # PIPA517500
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues surrounding cysteine 192 of human TRAF2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase TRAF2; E3 ubiquitin-protein ligase TRAF2 tumor necrosis factor type 2 receptor associated protein 3 tumor necrosis factor type 2 receptor-associated protein 3; RING-type E3 ubiquitin transferase TRAF2; TNF receptor-associated factor 2; tumor necrosis factor type 2 receptor associated protein 3; Tumor necrosis factor type 2 receptor-associated protein 3
Gene Aliases: AI325259; MGC:45012; TRAF2; TRAP; TRAP3
UniProt ID: (Human) Q12933, (Mouse) P39429
Entrez Gene ID: (Human) 7186, (Mouse) 22030
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.