Novus Biologicals
Manufacturer Code:NB10056715SS
Catalog # NB10056715T
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 33A1293 |
Immunogen | Anti-TRAF2 monoclonal antibody was raised against a fusion protein corresponding to amino acids 205 to 222 of human TRAF2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase TRAF2; E3 ubiquitin-protein ligase TRAF2 EC 6.3.2.- MGC:45012 TNF receptor-associated factor 2 TRAP3TRAP tumor necrosis factor type 2 receptor associated protein 3 Tumor necrosis factor type 2 receptor-associated protein 3; RING-type E3 ubiquitin transferase TRAF2; TNF receptor-associated factor 2; tumor necrosis factor type 2 receptor associated protein 3; Tumor necrosis factor type 2 receptor-associated protein 3
Gene Aliases: MGC:45012; TRAF2; TRAP; TRAP3
UniProt ID: (Human) Q12933
Entrez Gene ID: (Human) 7186
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.