Novus Biologicals
Manufacturer Code:NB10093535
Catalog # NB10093535
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-DIGSAPIVNAPEGNKVD corresponding to internal region according to Uniprot Zebrafish NP_571415.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 2 c-erbA-2 c-erbA-beta ERBA2MGC126109 ERBA-BETA GRTH MGC126110 NR1A2thyroid hormone receptor beta (avian erythroblastic leukemia viral (v-erb-a)oncogene homolog 2) Nuclear receptor subfamily 1 group A member 2 oncogene ERBA2 PRTH THR1pituitary resistance to thyroid hormone THRB1 THRB2 thyroid hormone receptor beta thyroid hormone receptor beta 1 thyroid hormone receptor beta (erythroblastic leukemia viral (v-erb-a)oncogene homolog 2 avian); c-erbA-2; c-erbA-beta; Nuclear receptor subfamily 1 group A member 2; oncogene ERBA2; thyroid hormone nuclear receptor beta variant 1; Thyroid hormone receptor beta; thyroid hormone receptor, beta (erythroblastic leukemia viral (v-erb-a) oncogene homolog 2, avian)
Gene Aliases: C-ERBA-2; C-ERBA-BETA; ERBA2; GRTH; NR1A2; PRTH; THR1; THRB; THRB1; THRB2
UniProt ID: (Human) P10828
Entrez Gene ID: (Human) 7068
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.