Proteintech
Manufacturer Code:108241AP150UL
Catalog # 50-172-6629
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TPT1 Fusion Protein Ag1268 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 21 kDa polypeptide; 21 kDa polypeptide under translational control; Fortilin; Fortilin Histamine releasing factor HRF p02 p23 TCTP TPT1; Histamine-releasing factor; HRF; Lens epithelial protein; p21; p23; TCTP; translationally regulated transcript (21 kDa); Translationally-controlled tumor protein
Gene Aliases: HRF; p02; p21; p23; TCTP; TPT1; Trt
UniProt ID: (Human) P13693, (Mouse) P63028, (Rat) P63029
Entrez Gene ID: (Human) 7178, (Mouse) 22070, (Rat) 116646
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.