Invitrogen
Manufacturer Code:PA5112777
Catalog # PIPA5112777
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Protein-tyrosine sulfotransferase 1 protein (283-370AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein-tyrosine sulfotransferase 1; Protein-tyrosine sulfotransferase 1 R75054 TANGO13A Tpst1 TPST-1 transport and golgi organization 13 homolog A Tyrosylprotein sulfotransferase 1 tyrosylprotein sulfotransferase-1; TPST-1; transport and golgi organization 13 homolog A; Tyrosylprotein sulfotransferase 1; tyrosylprotein sulfotransferase-1
Gene Aliases: R75054; TANGO13A; TPST1
UniProt ID: (Human) O60507, (Rat) Q3KR92, (Mouse) O70281
Entrez Gene ID: (Human) 8460, (Rat) 288617, (Mouse) 22021
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.