Proteintech
Manufacturer Code:128001AP150UL
Catalog # 50-172-8260
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TPRKB Fusion Protein Ag3551 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CGI 121 My019 PRPK binding protein TP53RK binding protein TPRKB; EKC/KEOPS complex subunit TPRKB; PRPK (p53-related protein kinase)-binding protein; PRPK-binding protein; TP53RK-binding protein
Gene Aliases: 0610033G21Rik; 1810034M08Rik; CGI-121; CGI121; DRWMS1; mDRWMS1; My019; RGD1309786; TPRKB
UniProt ID: (Human) Q9Y3C4, (Mouse) Q8QZZ7
Entrez Gene ID: (Human) 51002, (Mouse) 69786, (Rat) 297411
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.