Novus Biologicals
Manufacturer Code:NB1002867
Catalog # NB1002867
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 2300 and the C-terminus (residue 2349) of human Translocated Promoter Region Protein using the numbering given in entry NP_003283.1 (GeneID 7175). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Megator; NPC-associated intranuclear protein; nuclear pore complex-associated protein TPR; nuclear pore complex-associated protein TPR nucleoprotein TPR translocated promoter region (to activated MET oncogene) tumor potentiating region; Nucleoprotein TPR; translocated promoter region (to activated MET oncogene); Translocated promoter region protein; tumor potentiating region
Gene Aliases: TPR
UniProt ID: (Human) P12270
Entrez Gene ID: (Human) 7175
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.