Proteintech
Manufacturer Code:141201AP150UL
Catalog # 50-172-9225
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TPPII Fusion Protein Ag5296 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: TPP 2 TPP II TPP2 TPPII Tripeptidyl aminopeptidase Tripeptidyl peptidase 2 Tripeptidyl peptidase II; TPP-2; TPP-II; Tripeptidyl aminopeptidase; tripeptidyl peptidase II; Tripeptidyl-peptidase 2; Tripeptidyl-peptidase II
Gene Aliases: TPP-2; TPP2; TPPII
UniProt ID: (Human) P29144, (Mouse) Q64514
Entrez Gene ID: (Human) 7174, (Mouse) 22019
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.