Novus Biologicals
Manufacturer Code:NBP274570JF646
Catalog # N274570J646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2A2 |
Immunogen | Full length recombinant protein of human TPMT (NP_000358) produced in HEK293T cells. |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: EC 2.1.1.67 S-adenosyl-L-methionine:thiopurine S-methyltransferase Thiopurine methyltransferase thiopurine S-methyltransferase; S-adenosyl-L-methionine:thiopurine S-methyltransferase; Thiopurine methyltransferase; Thiopurine S-methyltransferase
Gene Aliases: TPMT; TPMTD
UniProt ID: (Human) P51580
Entrez Gene ID: (Human) 7172
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.