Invitrogen
Manufacturer Code:PA580176
Catalog # PIPA580176
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human TPMT recombit protein (Position: M1-K245). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: S-adenosyl-L-methionine:thiopurine S-methyltransferase; Thiopurine methyltransferase; Thiopurine S-methyltransferase; Thiopurine S-methyltransferase Thiopurine methyltransferase TPMT
Gene Aliases: AW106912; TPMT; TPMTD
UniProt ID: (Human) P51580, (Rat) Q9Z0T0, (Mouse) O55060
Entrez Gene ID: (Human) 7172, (Rat) 690050, (Mouse) 22017
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.