Proteintech
Manufacturer Code:110381AP150UL
Catalog # 50-172-6819
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TPM2 Fusion Protein Ag1508 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AMCD1 Beta tropomyosin DA1 DA2B TMSB TPM2 Tropomyosin 2 tropomyosin 2 (beta) Tropomyosin beta chain; Beta-tropomyosin; epididymis secretory protein Li 273; nemaline myopathy type 4; Tropomyosin beta chain; Tropomyosin-2
Gene Aliases: AMCD1; DA1; DA2B; HEL-S-273; NEM4; TMSB; Tpm-2; TPM2; Trop-2
UniProt ID: (Human) P07951, (Rat) P58775, (Mouse) P58774
Entrez Gene ID: (Human) 7169, (Rat) 500450, (Mouse) 22004
Molecular Function:
actin binding motor protein
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.