Invitrogen
Manufacturer Code:MA532982
Catalog # PIMA532982
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | TM31 |
Immunogen | Chicken gizzard tropomyosin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: alpha tropomyosin; alpha tropomyosin of brain; alpha tropomyosin of fibroblasts F2; alpha-Tm-1; alpha-Tm-2 isoform; Alpha-tropomyosin; alpha-tropomyosin (partial); alpha-tropomyosin 1; alpha-tropomyosin 2; alpha-tropomyosin of skeletal fast muscle; cardiomyopathy, hypertrophic 3; epididymis secretory protein Li 265; hepatoma alpha tropomyosin; sarcomeric tropomyosin kappa; smooth muscle alpha-tropomyosin; striated muscle alpha-tropomyosin; tropomyosin (CTm4); tropomyosin (CTm7); tropomyosin 1 (alpha); tropomyosin 3 alpha; Tropomyosin alpha-1 chain; Tropomyosin alpha-1 chain Alpha-tropomyosin Tropomyosin-1 Tpm1 Alpha-tm Tpma; Tropomyosin-1
Gene Aliases: AA986836; AI854628; alpha-fTM; Alpha-tm; C15orf13; CMD1Y; CMH3; HEL-S-265; HTM-alpha; LVNC9; TM2; Tm3; Tma2; Tmpa; TMSA; Tpm-1; TPM1; Tpma; tropomyosin-1
UniProt ID: (Human) P09493, (Chicken) P04268, (Rat) P04692, (Mouse) P58771
Entrez Gene ID: (Human) 7168, (Chicken) 396366, (Rat) 24851, (Mouse) 22003
Molecular Function: actin binding motor protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.