Invitrogen
Manufacturer Code:MA535706
Catalog # PIMA535706
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1283 |
Immunogen | A synthesized peptide derived from human Tropomyosin 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AA986836 AI854628 alpha-fTM Alpha-tm Alpha-tropomyosin BOS_10769 C15orf13 CMD1Y CMH3 HEL-S-265 HTM-alpha LVNC9 Tm1 TM2 Tm3 Tma2 Tmpa TMSA TPM1 Tpm-1 TPM1-1 TPM1-alpha TPMA Tropomyosin alpha-1 chain Tropomyosin-1 zgc:171719 zgc:55951 zgc:77009; alpha tropomyosin; Alpha-tropomyosin; hepatoma alpha tropomyosin; smooth muscle alpha-tropomyosin; striated muscle alpha-tropomyosin; tropomyosin 3 alpha; Tropomyosin alpha-1 chain; Tropomyosin-1
Gene Aliases: AA986836; AI854628; Alpha-tm; TM2; Tm3; Tma2; Tmpa; Tmsa; Tpm-1; Tpm1; Tpma
UniProt ID: (Rat) P04692, (Mouse) P58771
Entrez Gene ID: (Rat) 24851, (Mouse) 22003
Molecular Function: actin binding motor protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.