Invitrogen
Manufacturer Code:PA588151
Catalog # PIPA588151
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-245 of human TPM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha tropomyosin; Alpha-tropomyosin; cardiomyopathy, hypertrophic 3; epididymis secretory protein Li 265; hepatoma alpha tropomyosin; sarcomeric tropomyosin kappa; smooth muscle alpha-tropomyosin; striated muscle alpha-tropomyosin; tropomyosin 3 alpha; Tropomyosin alpha-1 chain; Tropomyosin-1
Gene Aliases: AA986836; AI854628; Alpha-tm; C15orf13; CMD1Y; CMH3; HEL-S-265; HTM-alpha; LVNC9; TM2; Tm3; Tma2; Tmpa; TMSA; Tpm-1; TPM1; Tpma
UniProt ID: (Human) P09493, (Rat) P04692, (Mouse) P58771
Entrez Gene ID: (Human) 7168, (Rat) 24851, (Mouse) 22003
Molecular Function: actin binding motor protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.