Invitrogen
Manufacturer Code:PA541996
Catalog # PIPA541996
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human TPM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha tropomyosin; Alpha-tropomyosin; cardiomyopathy, hypertrophic 3; epididymis secretory protein Li 265; hepatoma alpha tropomyosin; sarcomeric tropomyosin kappa; smooth muscle alpha-tropomyosin; striated muscle alpha-tropomyosin; tropomyosin 1 (alpha); Tropomyosin 1 (alpha) CMH3 TMSA TPM1-alpha CMD1Y C15orf13 HTM-alpha; tropomyosin 1 alpha chain; tropomyosin 1-1; tropomyosin 3 alpha; Tropomyosin alpha-1 chain; Tropomyosin-1; unnamed protein product
Gene Aliases: AA986836; AI854628; Alpha-tm; C15orf13; CMD1Y; CMH3; HEL-S-265; HTM-alpha; LVNC9; Tm1; TM2; Tm3; Tma2; Tmpa; TMSA; Tpm-1; TPM1; TPM1-1; TPMA; zgc:171719; zgc:55951; zgc:77009
UniProt ID: (Human) P09493, (Bovine) Q5KR49, (Rabbit) P58772, (Rat) P04692, (Mouse) P58771
Entrez Gene ID: (Horse) 100067330, (Human) 7168, (Dog) 478332, (Bovine) 281544, (Sheep) 100145865, (Rabbit) 100125989, (Rat) 24851, (Guinea pig) 100717160, (Mouse) 22003, (Zebrafish) 393908
Molecular Function: actin binding motor protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.