Invitrogen
Manufacturer Code:PA5102367
Catalog # PIPA5102367
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TPM1 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA986836 AI854628 alpha tropomyosin alpha-fTM Alpha-tm Alpha-tropomyosin alpha-tropomyosin 1 alpha-tropomyosin 2 alpha-tropomyosin of skeletal fast muscle C15orf13 cardiac alpha tropomyosin cardiomyopathy hypertrophic 3 CMD1Y CMH3 epididymis secretory protein Li 265 HEL-S-265 hepatoma alpha tropomyosin HTM-alpha LVNC9 sarcomeric tropomyosin kappa smooth muscle alpha-tropomyosin striated muscle alpha-tropomyosin Tm1 TM2 Tm3 Tma2 Tmpa TMSA Tpm1 Tpm-1 tpm1 protein TPM1-1 TPM1-alpha Tpma tropomyosin tropomyosin (CTm4) tropomyosin (CTm7) tropomyosin 1 tropomyosin 1 (alpha) tropomyosin 1 alpha tropomyosin 1-1 tropomyosin 3 alpha Tropomyosin alpha-1 chain Tropomyosin-1 zgc:171719 zgc:55951 zgc:77009; alpha tropomyosin; Alpha-tropomyosin; cardiomyopathy, hypertrophic 3; epididymis secretory protein Li 265; hepatoma alpha tropomyosin; sarcomeric tropomyosin kappa; smooth muscle alpha-tropomyosin; striated muscle alpha-tropomyosin; tropomyosin 3 alpha; Tropomyosin alpha-1 chain; Tropomyosin-1
Gene Aliases: AA986836; AI854628; Alpha-tm; C15orf13; CMD1Y; CMH3; HEL-S-265; HTM-alpha; LVNC9; TM2; Tm3; Tma2; Tmpa; TMSA; Tpm-1; TPM1; Tpma
UniProt ID: (Human) P09493, (Rat) P04692, (Mouse) P58771
Entrez Gene ID: (Human) 7168, (Rat) 24851, (Mouse) 22003
Molecular Function: actin binding motor protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.