Novus Biologicals
Manufacturer Code:NBP297926AF488
Catalog # NB103796
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human TPK1 fragment. |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.6.2 hTPK1 Placental protein 20 PP20thiamine kinase thiamin pyrophosphokinase 1 thiamine diphosphokinase Thiamine pyrophosphokinase 1; hTPK1; Placental protein 20; PP20; Thiamin pyrophosphokinase 1; thiamine diphosphokinase; thiamine kinase; Thiamine pyrophosphokinase 1
Gene Aliases: HTPK1; PP20; THMD5; TPK1
UniProt ID: (Human) Q9H3S4
Entrez Gene ID: (Human) 27010
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.