Proteintech
Manufacturer Code:109421AP150UL
Catalog # 50-172-6732
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TPK1 Fusion Protein Ag1404 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hTPK1; HTPK1 Placental protein 20 PP20 thiamin pyrophosphokinase 1 Thiamine pyrophosphokinase 1 TPK1; mTPK1; Placental protein 20; PP20; Thiamin pyrophosphokinase 1; thiamine diphosphokinase; thiamine kinase; thiamine pyrophosphokinase; Thiamine pyrophosphokinase 1
Gene Aliases: HTPK1; PP20; THMD5; TPK1
UniProt ID: (Human) Q9H3S4, (Mouse) Q9R0M5
Entrez Gene ID: (Human) 27010, (Rat) 680668, (Mouse) 29807
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.