Invitrogen
Manufacturer Code:MA518297
Catalog # PIMA518297
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | P3711 |
Immunogen | Recombit fragment corresponding to a region within amino acids 187 and 249 of Triosephosphate isomerase |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: epididymis secretory protein Li 49; Methylglyoxal synthase; TIM; TPI TIM triosephosphate isomerase 1 TPI1; Triose-phosphate isomerase; Triosephosphate isomerase
Gene Aliases: AI255506; HEL-S-49; TIM; TPI; Tpi-1; TPI1; TPID
UniProt ID: (Human) P60174, (Rat) P48500, (Mouse) P17751
Entrez Gene ID: (Human) 7167, (Rat) 24849, (Mouse) 21991
Molecular Function:
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.