Invitrogen
Manufacturer Code:PA599973
Catalog # PIPA599973
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TPI1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI255506 epididymis secretory protein Li 49 HEL-S-49 Methylglyoxal synthase MGC88108 TIM TPI TPI1 Tpi-1 TPID triosephosphate isomerase Triose-phosphate isomerase triosephosphate isomerase 1; epididymis secretory protein Li 49; Methylglyoxal synthase; TIM; Triose-phosphate isomerase; Triosephosphate isomerase
Gene Aliases: AI255506; HEL-S-49; TIM; TPI; Tpi-1; TPI1; TPID
UniProt ID: (Human) P60174, (Rat) P48500, (Mouse) P17751
Entrez Gene ID: (Human) 7167, (Rat) 24849, (Mouse) 21991
Molecular Function:
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.