Invitrogen
Manufacturer Code:PA590658
Catalog # PIPA590658
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-249 of human TPI1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymis secretory protein Li 49; Methylglyoxal synthase; TIM; Triose-phosphate isomerase; Triosephosphate isomerase
Gene Aliases: AI255506; HEL-S-49; TIM; TPI; Tpi-1; TPI1; TPID
UniProt ID: (Human) P60174, (Rat) P48500, (Mouse) P17751
Entrez Gene ID: (Human) 7167, (Rat) 24849, (Mouse) 21991
Molecular Function:
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.