Abnova
Manufacturer Code:PAB9644
Catalog # 89-134-297
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding S19 of rat Tph2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of Tph2.
For Research Use Only
Protein Aliases: Neuronal tryptophan hydroxylase; Ntph; Tryptophan 5-hydroxylase 2; Tryptophan 5-monooxygenase 2
Gene Aliases: Ntph; Tph2
UniProt ID: (Rat) Q8CGU9
Entrez Gene ID: (Rat) 317675
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.