Abnova
Manufacturer Code:PAB6927
Catalog # 89-132-806
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human TPH2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of TPH2.
For Research Use Only
Protein Aliases: FLJ37295; MGC138871; MGC138872; Neuronal tryptophan hydroxylase; NTPH; Tryptophan 5-hydroxylase 2; Tryptophan 5-monooxygenase 2
Gene Aliases: ADHD7; NTPH; TPH2
UniProt ID: (Human) Q8IWU9
Entrez Gene ID: (Human) 121278
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.