Invitrogen
Manufacturer Code:PA555498
Catalog # PIPA555498
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human TPCN2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Two pore calcium channel protein 2; two-pore calcium channel protein 2; Voltage-dependent calcium channel protein TPC2
Gene Aliases: SHEP10; TPC2; TPCN2
UniProt ID: (Human) Q8NHX9
Entrez Gene ID: (Human) 219931
Molecular Function: calcium channel ion channel sodium channel transporter voltage-gated calcium channel voltage-gated ion channel voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.