Invitrogen
Manufacturer Code:PA558915
Catalog # PIPA558915
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human TPCN1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Two pore calcium channel protein 1; Two pore segment channel 1 FLJ20612 KIAA1169 TPC1; two-pore channel 1; two-pore channel 1, homolog; two-pore segment channel 1; Voltage-dependent calcium channel protein TPC1
Gene Aliases: KIAA1169; TPC1; TPCN1
UniProt ID: (Human) Q9ULQ1
Entrez Gene ID: (Human) 53373
Molecular Function: calcium channel ion channel sodium channel transporter voltage-gated calcium channel voltage-gated ion channel voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.