Invitrogen
Manufacturer Code:PA541048
Catalog # PIPA541048
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human TPCN1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Two pore calcium channel protein 1; two pore channel 1; Two pore segment channel 1 FLJ20612 KIAA1169 TPC1; two-pore channel 1; two-pore channel 1, homolog; two-pore segment channel 1; Voltage-dependent calcium channel protein TPC1; voltage-gated Ca channel
Gene Aliases: 5730403B01Rik; KIAA1169; mKIAA1169; si:dkey-125i10.5; TPC1; TPCN1
UniProt ID: (Human) Q9ULQ1, (Mouse) Q9EQJ0, (Rat) Q9WTN5
Entrez Gene ID: (Rabbit) 100341301, (Human) 53373, (Dog) 477492, (Horse) 100051154, (Mouse) 252972, (Rat) 246215, (Guinea pig) 100717872, (Zebrafish) 567534
Molecular Function: calcium channel ion channel sodium channel transporter voltage-gated calcium channel voltage-gated ion channel voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.