Invitrogen
Manufacturer Code:PA571651
Catalog # PIPA571651
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 194-221 amino acids from the Central region of human TOR2A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Prosalusin; Salusin-alpha; Salusin-beta; Torsin family 2 member A; torsin family 2, member A; Torsin-2A; Torsin-2A Torsin family 2 member A Torsin-related protein 1 TOR2A TORP1; Torsin-related protein 1
Gene Aliases: HEMBA1005096; PSEC0218; TOR2A; TORP1; UNQ6408/PRO21181
UniProt ID: (Human) Q5JU69
Entrez Gene ID: (Human) 27433
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.