Proteintech
Manufacturer Code:195111AP150UL
Catalog # 50-173-2426
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Prosalusin; Prosalusin TOR2A TORP1 Torsin 2A Torsin family 2 member A torsin family 2 member A Torsin related protein 1; Salusin-alpha; Salusin-beta; Torsin family 2 member A; torsin family 2, member A; Torsin-2A; Torsin-related protein 1
Gene Aliases: HEMBA1005096; Prosalusin; PSEC0218; TOR2A; TORP1; UNQ6408/PRO21181
UniProt ID: (Human) Q5JU69, (Mouse) Q8R1J9
Entrez Gene ID: (Human) 27433, (Mouse) 30933
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.