Proteintech
Manufacturer Code:247691AP150UL
Catalog # 50-173-4530
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TOR1AIP2 Fusion Protein Ag20126 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 15 kDa interferon-responsive protein; IFRG15; interferon responsive gene 15; LULL1 Lumenal domain like LAP1 RP11 12M5.5 TOR1AIP2 torsin A interacting protein 2; lumenal domain like LAP1; Lumenal domain-like LAP1; torsin A interacting protein 2; Torsin-1A-interacting protein 2; Torsin-1A-interacting protein 2, isoform IFRG15
Gene Aliases: IFRG15; LULL1; NET9; TOR1AIP2
UniProt ID: (Human) Q8NFQ8
Entrez Gene ID: (Human) 163590
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.