Abnova
Manufacturer Code:H00163590K
Catalog # 89-207-565
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human TOR1AIP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human TOR1AIP2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: 15 kDa interferon-responsive protein; IFRG15; interferon responsive gene 15; LULL1 MGC126581 MGC138430 RP11-12M5.5; lumenal domain like LAP1; Lumenal domain-like LAP1; torsin A interacting protein 2; Torsin-1A-interacting protein 2; Torsin-1A-interacting protein 2, isoform IFRG15
Gene Aliases: IFRG15; LULL1; NET9; TOR1AIP2
UniProt ID: (Human) Q8NFQ8
Entrez Gene ID: (Human) 163590
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.