Novus Biologicals
Manufacturer Code:NBP122976
Catalog # NBP122976
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 995 and 1045 of human topoisomerase I binding arginine/serine-rich using the numbering given in entry topoisomerase I binding arginine/serine-rich NP_005793.2 (GeneID 10210). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase Topors; EC 6.3.2.- LUNtopoisomerase I binding arginine/serine-rich p53-binding protein 3 p53BP3 retinitis pigmentosa 31 (autosomal dominant) RP31 SUMO1-protein E3 ligase Topors topoisomerase I binding arginine/serine-rich E3 ubiquitin protein ligase Topoisomerase I-binding arginine/serine-rich protein Topoisomerase I-binding RING finger protein TP53BPLE3 ubiquitin-protein ligase Topors Tumor suppressor p53-binding protein 3; p53-binding protein 3; RING-type E3 ubiquitin transferase Topors; SUMO1-protein E3 ligase Topors; topoisomerase I binding, arginine/serine-rich, E3 ubiquitin protein ligase; Topoisomerase I-binding arginine/serine-rich protein; Topoisomerase I-binding RING finger protein; Tumor suppressor p53-binding protein 3
Gene Aliases: LUN; P53BP3; RP31; TOPORS; TP53BPL
UniProt ID: (Human) Q9NS56
Entrez Gene ID: (Human) 10210
Molecular Function:
isomerase
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.