Abnova
Manufacturer Code:H00010210A01
Catalog # 89-106-243
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 98 a.a. &bsim 205 a.a) partial recombinant protein with GST tag.,TOPORS (NP_005793 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant TOPORS.
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase Topors; LUN; p53-binding protein 3; P53BP3; RING-type E3 ubiquitin transferase Topors; RP31; SUMO1-protein E3 ligase Topors; topoisomerase I binding, arginine/serine-rich, E3 ubiquitin protein ligase; Topoisomerase I-binding arginine/serine-rich protein; Topoisomerase I-binding RING finger protein; TP53BPL; Tumor suppressor p53-binding protein 3
Gene Aliases: LUN; P53BP3; RP31; TOPORS; TP53BPL
UniProt ID: (Human) Q9NS56
Entrez Gene ID: (Human) 10210
Molecular Function:
isomerase
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.