Invitrogen
Manufacturer Code:PA575799
Catalog # PIPA575799
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human TOP3A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA topoisomerase 3-alpha; DNA topoisomerase 3-alpha DNA topoisomerase III alpha TOP3A TOP3; DNA topoisomerase III alpha; topo III-alpha; zinc finger, GRF-type containing 7
Gene Aliases: TOP3; TOP3A; ZGRF7
UniProt ID: (Human) Q13472
Entrez Gene ID: (Human) 7156
Molecular Function:
enzyme modulator
isomerase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.