Proteintech
Manufacturer Code:145251AP150UL
Catalog # 50-172-9515
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TOP3A Fusion Protein Ag6010 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA topoisomerase 3 alpha DNA topoisomerase III alpha TOP3 TOP3A topoisomerase (DNA) III alpha; DNA topoisomerase 3-alpha; DNA topoisomerase III alpha; Top IIIa; topo III-alpha; topoisomerase (DNA) 3 alpha; topoisomerase (DNA) III alpha; topoisomerase 3 alpha; zinc finger, GRF-type containing 7
Gene Aliases: TOP3; TOP3A; ZGRF7
UniProt ID: (Human) Q13472, (Mouse) O70157
Entrez Gene ID: (Human) 7156, (Rat) 303194, (Mouse) 21975
Molecular Function:
enzyme modulator
isomerase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.