Novus Biologicals
Manufacturer Code:NBP248472
Catalog # NBP248472
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | TOP1MT/488 |
Immunogen | Recombinant full-length human TOP1MT protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DNA topoisomerase I, mitochondrial; mitochondrial mitochondrial DNA topoisomerase I topoisomerase (DNA) I mitochondrial; mitochondrial topoisomerase IB; TOP1mt
Gene Aliases: TOP1MT
UniProt ID: (Human) Q86V82
Entrez Gene ID: (Human) 116447
Molecular Function: DNA binding protein DNA topoisomerase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.