OriGene
Manufacturer Code:TA809934
Catalog # 50-168-3108
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3B9 |
Immunogen | Human recombit protein fragment corresponding to amino acids 93-151 of human TOP1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DNA topoisomerase 1; DNA topoisomerase I; TOPI; type I DNA topoisomerase
Gene Aliases: TOP1; TOPI
UniProt ID: (Human) P11387
Entrez Gene ID: (Human) 7150
Molecular Function: DNA binding protein DNA topoisomerase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.