Invitrogen
Manufacturer Code:PA5106808
Catalog # PIPA5106808
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TOP1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ab2-086 AI467334 D130064I21Rik DNA topoisomerase 1 DNA topoisomerase I RP3-511B24.2 TOP1 Top-1 TOPI topoisomerase (DNA) I type I DNA topoisomerase; DNA topoisomerase 1; DNA topoisomerase I; type I DNA topoisomerase
Gene Aliases: Ab2-086; AI467334; D130064I21Rik; Top-1; TOP1; TOPI
UniProt ID: (Human) P11387, (Rat) Q9WUL0, (Mouse) Q04750
Entrez Gene ID: (Human) 7150, (Rat) 64550, (Mouse) 21969
Molecular Function:
DNA binding protein
DNA topoisomerase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.