Proteintech
Manufacturer Code:150711AP150UL
Catalog # 50-172-9951
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TOMM7 Fusion Protein Ag7105 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6.2 kd protein; Mitochondrial import receptor subunit TOM7 homolog; TOM7 TOMM07 TOMM7; Translocase of outer membrane 7 kDa subunit homolog; translocase of outer mitochondrial membrane 7 homolog
Gene Aliases: 1110020J08Rik; AD-014; AW047273; TOM7; TOMM07; TOMM7
UniProt ID: (Human) Q9P0U1, (Mouse) Q9D173
Entrez Gene ID: (Human) 54543, (Mouse) 66169, (Rat) 685620
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.