Novus Biologicals
Manufacturer Code:NBP294075100UL
Catalog # NB022709
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-90 of human TOMM40 (NP_001122389.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C19orf1mitochondrial import receptor subunit TOM40 homolog D19S1177E mitochondrial outer membrane protein PER-EC1 PEREC1Translocase of outer membrane 40 kDa subunit homolog Protein Haymaker TOM40p38.5 translocase of outer mitochondrial membrane 40 homolog (yeast); Mitochondrial import receptor subunit TOM40 homolog; mitochondrial outer membrane protein; p38.5; Protein Haymaker; Translocase of outer membrane 40 kDa subunit homolog; translocase of outer mitochondrial membrane 40 homolog
Gene Aliases: C19orf1; D19S1177E; PER-EC1; PEREC1; TOM40; TOMM40
UniProt ID: (Human) O96008
Entrez Gene ID: (Human) 10452
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.