Invitrogen
Manufacturer Code:PA586739
Catalog # PIPA586739
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 57-102 of Human TOMM20. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Mitochondrial 20 kDa outer membrane protein; Mitochondrial import receptor subunit TOM20 homolog; Mitochondrial import receptor subunit TOM20 homolog Mitochondrial 20 kDa outer membrane proteinOuter mitochondrial membrane receptor Tom20 TOM20 KIAA0016; outer mitochondrial membrane receptor rTOM20; Outer mitochondrial membrane receptor Tom20; translocase of outer mitochondrial membrane 20 homolog type II
Gene Aliases: 1810060K07Rik; BB284719; Gm19268; KIAA0016; MAS20; mKIAA0016; MOM19; TOM20; TOMM20
UniProt ID: (Human) Q15388, (Rat) Q62760, (Mouse) Q9DCC8
Entrez Gene ID: (Human) 9804, (Rat) 266601, (Mouse) 67952
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.