Novus Biologicals
Manufacturer Code:NBP303878100UL
Catalog # NB092663
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human TOMM20. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA0016Outer mitochondrial membrane receptor Tom20 MAS20 MGC117367 Mitochondrial 20 kDa outer membrane protein mitochondrial import receptor subunit TOM20 homolog MOM19 TOM20 translocase of outer mitochondrial membrane 20 homolog (yeast) translocase of outer mitochondrial membrane 20 homolog type II; Mitochondrial 20 kDa outer membrane protein; Mitochondrial import receptor subunit TOM20 homolog; Outer mitochondrial membrane receptor Tom20; translocase of outer mitochondrial membrane 20 homolog type II
Gene Aliases: KIAA0016; MAS20; MOM19; TOM20; TOMM20
UniProt ID: (Human) Q15388
Entrez Gene ID: (Human) 9804
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.